Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Methoxybiphenyl-4-sulfonyl chloride, 95%, Acros Organics

Click to view available options
Quantity:
1g
5g
186550-26-5
C13H11ClO3S
282.75
DVQBUJTYQXKLKZ-UHFFFAOYSA-N
3'-methoxy-biphenyl-4-sulfonyl chloride,3'-methoxybiphenyl-4-sulfonyl chloride,3'-methoxy 1,1'-biphenyl-4-sulfonyl chloride,3'-methoxy-1,1'-biphenyl-4-sulfonyl chloride,1,1'-biphenyl-4-sulfonylchloride, 3'-methoxy,4-3-methoxyphenyl benzenesulfonyl chloride,3'-methoxy-biphenyl-4-sulfonylchloride,3/'-methoxybiphenyl-4-sulfonyl chloride
10826798
4-(3-methoxyphenyl)benzenesulfonyl chloride
COC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
186550-26-5
C13H11ClO3S
282.75
DVQBUJTYQXKLKZ-UHFFFAOYSA-N
3'-methoxy-biphenyl-4-sulfonyl chloride,3'-methoxybiphenyl-4-sulfonyl chloride,3'-methoxy 1,1'-biphenyl-4-sulfonyl chloride,3'-methoxy-1,1'-biphenyl-4-sulfonyl chloride,1,1'-biphenyl-4-sulfonylchloride, 3'-methoxy,4-3-methoxyphenyl benzenesulfonyl chloride,3'-methoxy-biphenyl-4-sulfonylchloride,3/'-methoxybiphenyl-4-sulfonyl chloride
10826798
4-(3-methoxyphenyl)benzenesulfonyl chloride
COC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
Melting Point 81°C to 83°C
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 94% min. (HPLC)
Formula Weight 282.75
Percent Purity 95%
Chemical Name or Material 3'-Methoxybiphenyl-4-sulfonyl chloride

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS Signal Word: Danger