Learn More
3-(Methoxycarbonyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals
Description
Reagent used for tandem-type Pd(II)-catalyzed oxidative Heck reaction and intramolecular C-H amidation sequence, copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides, one-pot ipso-nitration of arylboronic acids, copper-catalyzed nitration, cyclocondensation followed by palladium-phosphine-catalyzed Suzuki-Miyaura coupling. Reagent used in Preparation of biaryls via nickel-catalyzed Suzuki-Miyaura cross-coupling reaction of aryl halides with arylboronic acid6, chromenones and their bradykinin B1 antagonistic active, Pt nanoparticles at Photoactive metal-organic frameworks resulting in efficient hydrogen evolution via synergistic photoexcitation and electron injecti, salicylate-based thienylbenzoic acids as E. coli methionine aminopeptidase inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reagent used for tandem-type Pd(II)-catalyzed oxidative Heck reaction and intramolecular C-H amidation sequence, copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides, one-pot ipso-nitration of arylboronic acids, copper-catalyzed nitration, cyclocondensation followed by palladium-phosphine-catalyzed Suzuki-Miyaura coupling. Reagent used in Preparation of biaryls via nickel-catalyzed Suzuki-Miyaura cross-coupling reaction of aryl halides with arylboronic acid6, chromenones and their bradykinin B1 antagonistic active, Pt nanoparticles at Photoactive metal-organic frameworks resulting in efficient hydrogen evolution via synergistic photoexcitation and electron injecti, salicylate-based thienylbenzoic acids as E. coli methionine aminopeptidase inhibitor.
Solubility
Reacts with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 136°C |
Quantity | 5 g |
Solubility Information | Reacts with water. |
IUPAC Name | (3-methoxycarbonylphenyl)boronic acid |
Formula Weight | 179.97 |
Percent Purity | 97% |
Chemical Name or Material | 3-(Methoxycarbonyl)benzeneboronic acid |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.