Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxycarbonylphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
99769-19-4
C8H9BO4
179.97
MFCD02093046
ALTLCJHSJMGSLT-UHFFFAOYSA-N
3-methoxycarbonyl phenylboronic acid,3-methoxycarbonylphenyl boronic acid,3-methoxycarbonyl phenyl boronic acid,3-methoxycarbonyl benzeneboronic acid,3-methoxycarbonylphenylbaronic acid,methyl 3-dihydroxyboranyl benzoate,m-methoxycarbonyl phenylboronic acid,3-carbomethoxy-phenylboronic acid
2734714
(3-methoxycarbonylphenyl)boronic acid
B(C1=CC(=CC=C1)C(=O)OC)(O)O
99769-19-4
C8H9BO4
179.97
MFCD02093046
ALTLCJHSJMGSLT-UHFFFAOYSA-N
3-methoxycarbonyl phenylboronic acid,3-methoxycarbonylphenyl boronic acid,3-methoxycarbonyl phenyl boronic acid,3-methoxycarbonyl benzeneboronic acid,3-methoxycarbonylphenylbaronic acid,methyl 3-dihydroxyboranyl benzoate,m-methoxycarbonyl phenylboronic acid,3-carbomethoxy-phenylboronic acid
2734714
(3-methoxycarbonylphenyl)boronic acid
B(C1=CC(=CC=C1)C(=O)OC)(O)O
Melting Point 187°C to 191°C
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 179.97
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 3-Methoxycarbonylphenylboronic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning