Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-3-Methoxymorphinan hydrochloride, For drug analysis, MilliporeSigma™ Supelco™

$290.00 - $988.00

Specifications

Grade Analytical Standard (for drug analysis)
Linear Formula C17H23 NO · HCl
Physical Form White Powder
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-2434
View Documents
MilliporeSigma Supelco
M18725MG
25 mg
Each for $290.00
Add to cart
 
11-101-2433
View Documents
MilliporeSigma Supelco
M187100MG
100 mg
Each for $988.00
Add to cart
 
Description

Description

Featured Industries: Pharmaceutical (small molecule)
Specifications

Specifications

Analytical Standard (for drug analysis)
C17H23 NO · HCl
White Powder
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit