Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxyphenethyl alcohol, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
5020-41-7
C9H12O2
152.19
MFCD00002893
UPPGEJSCUZMCMW-UHFFFAOYSA-N
2-3-methoxyphenyl ethanol,3-methoxyphenethyl alcohol,benzeneethanol, 3-methoxy,2-3-methoxyphenyl ethan-1-ol,phenethyl alcohol, m-methoxy,3-methoxybenzeneethanol,m-methoxyphenethyl alcohol,1-2-hydroxyethyl-3-methoxybenzene,acmc-20abd2,3-methoxyphenethylalcohol
78724
2-(3-methoxyphenyl)ethanol
COC1=CC=CC(=C1)CCO
5020-41-7
C9H12O2
152.19
MFCD00002893
UPPGEJSCUZMCMW-UHFFFAOYSA-N
2-3-methoxyphenyl ethanol,3-methoxyphenethyl alcohol,benzeneethanol, 3-methoxy,2-3-methoxyphenyl ethan-1-ol,phenethyl alcohol, m-methoxy,3-methoxybenzeneethanol,m-methoxyphenethyl alcohol,1-2-hydroxyethyl-3-methoxybenzene,acmc-20abd2,3-methoxyphenethylalcohol
78724
2-(3-methoxyphenyl)ethanol
COC1=CC=CC(=C1)CCO
Boiling Point 141.0°C to 143.0°C (12.0mmHg)
Density 1.0750g/mL
Specific Gravity 1.075
Flash Point >113°C
Packaging Glass bottle
Refractive Index 1.5381
NMR Consistent with structure
Quantity 25g
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3OC6H4CH2CH2OH
Formula Weight 152.19
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-Methoxyphenethyl alcohol
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-705-2