Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Methoxyphenyl)acetonitrile, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
Packaging:
Glass bottle
19924-43-7
C9H9NO
147.18
MFCD00001910
LXKNAUOWEJWGTE-UHFFFAOYSA-N
3-methoxyphenyl acetonitrile, 2-3-methoxyphenyl acetonitrile, 3-methoxyphenylacetonitrile, 3-methoxybenzyl cyanide, m-methoxyphenyl acetonitrile, 3-cyanomethyl anisole, benzeneacetonitrile, 3-methoxy, 3-methoxybenzeneacetonitrile, m-methoxybenzyl cyanide, 2-3-methoxyphenyl ethanenitrile
88310
2-(3-methoxyphenyl)acetonitrile
COC1=CC=CC(=C1)CC#N
19924-43-7
C9H9NO
147.18
MFCD00001910
LXKNAUOWEJWGTE-UHFFFAOYSA-N
3-methoxyphenyl acetonitrile, 2-3-methoxyphenyl acetonitrile, 3-methoxyphenylacetonitrile, 3-methoxybenzyl cyanide, m-methoxyphenyl acetonitrile, 3-cyanomethyl anisole, benzeneacetonitrile, 3-methoxy, 3-methoxybenzeneacetonitrile, m-methoxybenzyl cyanide, 2-3-methoxyphenyl ethanenitrile
88310
2-(3-methoxyphenyl)acetonitrile
COC1=CC=CC(=C1)CC#N
Specific Gravity 1.05
Flash Point 98°C
Boiling Point 164°C to 165°C (20.0mmHg)
Refractive Index 1.5297 to 1.5317
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Density 1.0500g/mL
Quantity 10g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3OC6H4CH2CN
Beilstein 10, III, 429
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene
Formula Weight 147.18
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material (3-Methoxyphenyl)acetonitrile, 97%
Show More Show Less

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for

GHS Signal Word: Danger

EINECSNumber : 243-428-5

TSCA : TSCA