Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Methoxyphenyl isocyanate, 99%

Click to view available options
Quantity:
5g
25g
100g
18908-07-1
C8H7NO2
149.149
MFCD00002019
NPOVTGVGOBJZPY-UHFFFAOYSA-N
3-methoxyphenyl isocyanate,benzene, 1-isocyanato-3-methoxy,isocyanic acid 3-methoxyphenyl ester,3-isocyanatoanisole,3-methoxyphenylisocyanate,m-methoxyphenyl isocyanate,1-isocyanato-3-methoxy-benzene,3-methoxybenzenisocyanate,m-anisyl isocyanate,m-methoxyphenylisocyanate
87843
1-isocyanato-3-methoxybenzene
COC1=CC=CC(=C1)N=C=O
18908-07-1
C8H7NO2
149.149
MFCD00002019
NPOVTGVGOBJZPY-UHFFFAOYSA-N
3-methoxyphenyl isocyanate,benzene, 1-isocyanato-3-methoxy,isocyanic acid 3-methoxyphenyl ester,3-isocyanatoanisole,3-methoxyphenylisocyanate,m-methoxyphenyl isocyanate,1-isocyanato-3-methoxy-benzene,3-methoxybenzenisocyanate,m-anisyl isocyanate,m-methoxyphenylisocyanate
87843
1-isocyanato-3-methoxybenzene
COC1=CC=CC(=C1)N=C=O
Flash Point 96°C (204°F)
Boiling Point 94°C to 95°C (10mmHg)
Quantity 100g
Odor Pungent
UN Number UN2206
Beilstein 775986
Formula Weight 149.15
Percent Purity 99%
Chemical Name or Material 3-Methoxyphenyl isocyanate

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

TSCA : TSCA