Alfa Aesar™ 3-Methoxyphenyl isocyanate, 99%

Manufacturer: Alfa Aesar™ B2432914

Quantity 25g

 View more versions of this product

Catalog No. AAB2432914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18908-07-1
    Molecular Formula C8H7NO2
    Formula Weight 149.15
    MDL Number MFCD00002019
    UN Number UN2206
    Beilstein 775986
    Synonym m-Anisyl isocyanate
    Chemical Name or Material 3-Methoxyphenyl isocyanate

    Specifications

    Formula Weight 149.15
    MDL Number MFCD00002019
    UN Number UN2206
    Synonym m-Anisyl isocyanate
    Quantity 25g
    Odor Pungent
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H334-H302-H315-H319-H335
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    Molecular Formula C8H7NO2
    Boiling Point 94° to 95°C (10mmHg)
    CAS 18908-07-1
    Beilstein 775986
    Flash Point 96°C (204°F)
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
    Chemical Name or Material 3-Methoxyphenyl isocyanate
    Assay 99%

    GHS H Statement
    H331-H334-H302-H315-H319-H335
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P284a-P261-P280-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit