Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxyphenylacetic acid, 99.5%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1798-09-0
C9H10O3
166.18
MFCD00004334
LEGPZHPSIPPYIO-UHFFFAOYSA-N
3-methoxyphenylacetic acid,2-3-methoxyphenyl acetic acid,m-methoxyphenylacetic acid,3-methoxyphenyl acetic acid,3-methoxybenzeneacetic acid,benzeneacetic acid, 3-methoxy,acetic acid, m-methoxyphenyl,unii-25xlo0t6my,3-methoxy-phenyl-acetic acid,25xlo0t6my
15719
2-(3-methoxyphenyl)acetic acid
COC1=CC=CC(=C1)CC(=O)O
1798-09-0
C9H10O3
166.18
MFCD00004334
LEGPZHPSIPPYIO-UHFFFAOYSA-N
3-methoxyphenylacetic acid,2-3-methoxyphenyl acetic acid,m-methoxyphenylacetic acid,3-methoxyphenyl acetic acid,3-methoxybenzeneacetic acid,benzeneacetic acid, 3-methoxy,acetic acid, m-methoxyphenyl,unii-25xlo0t6my,3-methoxy-phenyl-acetic acid,25xlo0t6my
15719
2-(3-methoxyphenyl)acetic acid
COC1=CC=CC(=C1)CC(=O)O
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Color White to Yellow
Melting Point 71.0°C to 73.0°C
CAS Min % 99.4
CAS Max % 100.0
Assay Percent Range 99.50%
Linear Formula CH3OC6H4CH2CO2H
Beilstein 10, I, 82
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol
Formula Weight 166.18
Physical Form Flakes
Percent Purity 99.5%
Chemical Name or Material 3-Methoxyphenylacetic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 217-282-8