Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxyphenylmagnesium bromide, 1.0 M in 2-MeTHF, Thermo Scientific Chemicals

Catalog Number AAH54780AE
Click to view available options
Quantity:
100 mL
36282-40-3
C7H7BrMgO
211.34
MFCD00672002
AJKNYEHIJAMGHX-UHFFFAOYSA-M
3-methoxyphenylmagnesium bromide, 3-methoxyphenyl magnesium bromide, 3-methoxyphenylmagnesium bromide solution, 3-methoxyphenylmagnesiumbromide, 3-methoxyphenylmagnesiumbromid, bromo 3-methoxyphenyl magnesium, 3-methoxy phenylmagnesium bromide, 3-methoxyphenyl-magnesium bromide, m-methoxyphenyl magnesium bromide, 3-methoxylphenyl magnesium bromide, grignard reagent
4605969
magnesium(2+) anisole bromide
COC1=CC=CC([Mg]Br)=C1
36282-40-3
C7H7BrMgO
211.34
MFCD00672002
AJKNYEHIJAMGHX-UHFFFAOYSA-M
3-methoxyphenylmagnesium bromide, 3-methoxyphenyl magnesium bromide, 3-methoxyphenylmagnesium bromide solution, 3-methoxyphenylmagnesiumbromide, 3-methoxyphenylmagnesiumbromid, bromo 3-methoxyphenyl magnesium, 3-methoxy phenylmagnesium bromide, 3-methoxyphenyl-magnesium bromide, m-methoxyphenyl magnesium bromide, 3-methoxylphenyl magnesium bromide, grignard reagent
4605969
magnesium(2+) anisole bromide
COC1=CC=CC([Mg]Br)=C1

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Formula Weight 211.34
Concentration or Composition (by Analyte or Components) 1.0M in 2-MeTHF
Chemical Name or Material 3-Methoxyphenylmagnesium bromide
Hazard Category H225-H302-H314-H335
Hazard Statement GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit