Alfa Aesar™ 3-Methoxyquinoline-5-boronic acid pinacol ester, 96%

Manufacturer: Alfa Aesar™ H54211MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH54211MD

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C16H20BNO3
    Formula Weight 285.15
    MDL Number MFCD20527212
    Chemical Name or Material 3-Methoxyquinoline-5-boronic acid pinacol ester

    Specifications

    Formula Weight 285.15
    Molecular Formula C16H20BNO3
    MDL Number MFCD20527212
    Quantity 250mg
    Chemical Name or Material 3-Methoxyquinoline-5-boronic acid pinacol ester
    Assay 96%

    Description

    Applications
    3-Methoxyquinoline-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit