Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxysalicylic acid, 97%, Acros Organics

Click to view available options
Quantity:
25g
877-22-5
C8H8O4
168.148
MFCD00002445
AUZQQIPZESHNMG-UHFFFAOYSA-N
3-methoxysalicylic acid,o-vanillic acid,acide orthovanillique,benzoic acid, 2-hydroxy-3-methoxy,m-anisic acid, 2-hydroxy,3-hydroxy-m-anisic acid,unii-3j31y0e2im,2-hydroxy-3-methoxy-benzoic acid,3-methoxysalicyclic acid,2-hydroxy-3-methoxy benzoic acid
70140
CHEBI:68496
2-hydroxy-3-methoxybenzoic acid
COC1=CC=CC(=C1O)C(=O)O
877-22-5
C8H8O4
168.148
MFCD00002445
AUZQQIPZESHNMG-UHFFFAOYSA-N
3-methoxysalicylic acid,o-vanillic acid,acide orthovanillique,benzoic acid, 2-hydroxy-3-methoxy,m-anisic acid, 2-hydroxy,3-hydroxy-m-anisic acid,unii-3j31y0e2im,2-hydroxy-3-methoxy-benzoic acid,3-methoxysalicyclic acid,2-hydroxy-3-methoxy benzoic acid
70140
CHEBI:68496
2-hydroxy-3-methoxybenzoic acid
COC1=CC=CC(=C1O)C(=O)O
Melting Point 147°C to 152°C
Infrared Spectrum Authentic
Quantity 25g
Color Beige
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3OC6H3(OH)CO2H
Beilstein 10, 376
Formula Weight 168.15
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 3-Methoxysalicylic acid, 97%
Show More Show Less