Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Methoxythiophene, 99%

Click to view available options
Quantity:
1g
5g
25g
17573-92-1
C5H6OS
114.162
MFCD00043894
RFSKGCVUDQRZSD-UHFFFAOYSA-N
thiophene, 3-methoxy,3-methoxy-thiophene,pubchem8863,acmc-1bow6,3-methoxythiophene,ksc491i4p,bidd:gt0411
140249
3-methoxythiophene
COC1=CSC=C1
17573-92-1
C5H6OS
114.162
MFCD00043894
RFSKGCVUDQRZSD-UHFFFAOYSA-N
thiophene, 3-methoxy,3-methoxy-thiophene,pubchem8863,acmc-1bow6,3-methoxythiophene,ksc491i4p,bidd:gt0411
140249
3-methoxythiophene
COC1=CSC=C1
Flash Point 49°C (120°F)
Odor Recognizable
Boiling Point 60°C to 61°C (12mmHg)
Quantity 1g
UN Number UN3271
Beilstein 106404
Formula Weight 114.17
Percent Purity 99%
Chemical Name or Material 3-Methoxythiophene

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.