Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxythiophene, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
17573-92-1
C5H6OS
114.17
MFCD00043894
RFSKGCVUDQRZSD-UHFFFAOYSA-N
thiophene, 3-methoxy,3-methoxy-thiophene,pubchem8863,acmc-1bow6,3-methoxythiophene,ksc491i4p,bidd:gt0411
140249
3-methoxythiophene
COC1=CSC=C1
17573-92-1
C5H6OS
114.17
MFCD00043894
RFSKGCVUDQRZSD-UHFFFAOYSA-N
thiophene, 3-methoxy,3-methoxy-thiophene,pubchem8863,acmc-1bow6,3-methoxythiophene,ksc491i4p,bidd:gt0411
140249
3-methoxythiophene
COC1=CSC=C1
Density 1.1430g/mL
Boiling Point 80.0°C to 82.0°C (65.0mmHg)
Flash Point 49°C
Packaging Glass bottle
Color Brown
Refractive Index 1.5300 to 1.5320 (20°C, 589nm)
Quantity 1g
Specific Gravity 1.143
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 114.17
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Methoxythiophene
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning