Alfa Aesar™ 3-Methoxythiophenol, 98%

Manufacturer: Alfa Aesar™ A1064814

Quantity 25g

 View more versions of this product

Catalog No. AAA1064814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15570-12-4
    Molecular Formula C7H8OS
    Formula Weight 140.21
    MDL Number MFCD00004841
    Beilstein 2041496
    Synonym 3-Mercaptoanisole; 3-Methoxybenzenethiol
    Chemical Name or Material 3-Methoxythiophenol

    Specifications

    Formula Weight 140.21
    Health Hazard 3 GHS P Statement
    P261-P280h-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Beilstein 2041496
    Synonym 3-Mercaptoanisole; 3-Methoxybenzenethiol
    Flash Point 96°C (204°F)
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C7H8OS
    Boiling Point 225° to 227°C
    Quantity 25g
    Odor Unpleasant
    MDL Number MFCD00004841
    CAS 15570-12-4
    Health Hazard 1 Warning
    Chemical Name or Material 3-Methoxythiophenol
    Assay 98%

    Description

    Applications
    3-Methoxythiophenol is used in the preparation of allenes.

    Solubility
    Not miscible or difficult to mix in water. Soluble in methanol, benzene, hexane, toluene and dichloromethane.

    Notes
    Store under dry inert gas. It is sensitive to air. Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280h-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit