Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1,2-cyclopentanedione, 99%, Thermo Scientific Chemicals

Catalog Number AC176460100
Click to view available options
Quantity:
10 g
50 g
250 g
765-70-8
C6H8O2
112.13
MFCD00001417
OACYKCIZDVVNJL-UHFFFAOYSA-N
3-methyl-1,2-cyclopentanedione, 1,2-cyclopentanedione, 3-methyl, methylcyclopentenolone diketo form, fema no. 2700, methylcyclopentenolone natural, kentonarome, benzil-related compound, 45, 3-methyl-1,2-cyclopentanedione maple lactone, maple lactone mcp, methyl-cyclopentenolone
61209
3-methylcyclopentane-1,2-dione
CC1CCC(=O)C1=O
765-70-8
C6H8O2
112.13
MFCD00001417
OACYKCIZDVVNJL-UHFFFAOYSA-N
3-methyl-1,2-cyclopentanedione, 1,2-cyclopentanedione, 3-methyl, methylcyclopentenolone diketo form, fema no. 2700, methylcyclopentenolone natural, kentonarome, benzil-related compound, 45, 3-methyl-1,2-cyclopentanedione maple lactone, maple lactone mcp, methyl-cyclopentenolone
61209
3-methylcyclopentane-1,2-dione
CC1CCC(=O)C1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 105.0°C to 110.0°C
Color White
Density 100g/mL
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 07, I, 310
Solubility Information Solubility in water: soluble.
Formula Weight 112.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Methyl-1,2-cyclopentanedione
Show More Show Less
EINECSNumber 212-154-8

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit