Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1,2-cyclopentanedione, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
765-70-8
C6H8O2
112.13
MFCD00001417
OACYKCIZDVVNJL-UHFFFAOYSA-N
3-methyl-1,2-cyclopentanedione, 1,2-cyclopentanedione, 3-methyl, methylcyclopentenolone diketo form, fema no. 2700, methylcyclopentenolone natural, kentonarome, benzil-related compound, 45, 3-methyl-1,2-cyclopentanedione maple lactone, maple lactone mcp, methyl-cyclopentenolone
61209
3-methylcyclopentane-1,2-dione
CC1CCC(=O)C1=O
765-70-8
C6H8O2
112.13
MFCD00001417
OACYKCIZDVVNJL-UHFFFAOYSA-N
3-methyl-1,2-cyclopentanedione, 1,2-cyclopentanedione, 3-methyl, methylcyclopentenolone diketo form, fema no. 2700, methylcyclopentenolone natural, kentonarome, benzil-related compound, 45, 3-methyl-1,2-cyclopentanedione maple lactone, maple lactone mcp, methyl-cyclopentenolone
61209
3-methylcyclopentane-1,2-dione
CC1CCC(=O)C1=O
Quantity 250g
Melting Point 105.0°C to 110.0°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Density 100g/mL
Flash Point 100°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Beilstein 07, I, 310
Solubility Information Solubility in water: soluble.
Formula Weight 112.13
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 3-Methyl-1,2-cyclopentanedione
Show More Show Less

EINECSNumber : 212-154-8