Alfa Aesar™ 3-Methyl-1-(2-phenylethyl)-4-piperidone, 97%

Manufacturer: Alfa Aesar™ H2753203

Quantity 1g

 View more versions of this product

Catalog No. AAH2753203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 129164-39-2
    Molecular Formula C14H19NO
    Formula Weight 217.32
    MDL Number MFCD01699280
    Chemical Name or Material 3-Methyl-1-(2-phenylethyl)-4-piperidone

    Specifications

    Formula Weight 217.32
    Molecular Formula C14H19NO
    MDL Number MFCD01699280
    CAS 129164-39-2
    Quantity 1g
    Chemical Name or Material 3-Methyl-1-(2-phenylethyl)-4-piperidone
    Assay 97%

    Description

    Applications
    As intermediates in the synthesis of several piperidine derivatives. 4-Piperidone is used as an intermediate in the manufacture of chemicals and pharmaceutical drugs.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit