Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1,3-butanediol, 98%, ACROS Organics™

Click to view available options
Quantity:
1L
Packaging:
Glass bottle
2568-33-4
C5H12O2
104.149
MFCD00059655
XPFCZYUVICHKDS-UHFFFAOYSA-N
3-methyl-1,3-butanediol,1,3-butanediol, 3-methyl,isopentyldiol,2-methyl-2,4-butanediol,unii-19nol5474q,2,4-dihydroxy-2-methylbutane,3-methyl-butane-1,3-diol,1,3-butanediol,3-methyl,3-methyl-1,3-butandiol,3-methylbutan-1,3-diol
247470
3-methylbutane-1,3-diol
CC(C)(CCO)O
2568-33-4
C5H12O2
104.149
MFCD00059655
XPFCZYUVICHKDS-UHFFFAOYSA-N
3-methyl-1,3-butanediol,1,3-butanediol, 3-methyl,isopentyldiol,2-methyl-2,4-butanediol,unii-19nol5474q,2,4-dihydroxy-2-methylbutane,3-methyl-butane-1,3-diol,1,3-butanediol,3-methyl,3-methyl-1,3-butandiol,3-methylbutan-1,3-diol
247470
3-methylbutane-1,3-diol
CC(C)(CCO)O
Packaging Glass bottle
Color Colorless
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Quantity 1L
Infrared Spectrum Authentic
Density 0.9760g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3)2C(OH)CH2CH2OH
Formula Weight 104.15
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 3-Methyl-1, 3-butanediol
Show More Show Less