Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-butanethiol 96%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
541-31-1
C5H12S
104.211
MFCD00004899
GIJGXNFNUUFEGH-UHFFFAOYSA-N
isoamyl mercaptan,3-methyl-1-butanethiol,1-butanethiol, 3-methyl,isopentyl mercaptan,3-methylbutanethiol,isoamylthiol,isopentanethiol,isoamylmercaptan,unii-mmk4sun45e,isoamyl sulfhydrate
10925
3-methylbutane-1-thiol
CC(C)CCS
541-31-1
C5H12S
104.211
MFCD00004899
GIJGXNFNUUFEGH-UHFFFAOYSA-N
isoamyl mercaptan,3-methyl-1-butanethiol,1-butanethiol, 3-methyl,isopentyl mercaptan,3-methylbutanethiol,isoamylthiol,isopentanethiol,isoamylmercaptan,unii-mmk4sun45e,isoamyl sulfhydrate
10925
3-methylbutane-1-thiol
CC(C)CCS
Density 0.835g/mL
Refractive Index 1.4420 to 1.4460 (20°C, 589nm)
Quantity 25g
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula (CH3)2CHCH2CH2SH
Beilstein 01, 405
Merck Index 09, 4970
Formula Weight 104.21
Physical Form Liquid
Percent Purity ≥95%
Chemical Name or Material 3-Methyl-1-butanethiol, 96%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-774-3

TSCA : TSCA