Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-butanol, 99%, anhydrous, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
1L
Packaging:
AcroSeal™ Glass Bottle
123-51-3
C5H12O
88.15
PHTQWCKDNZKARW-UHFFFAOYSA-N
3-methyl-1-butanol,isoamyl alcohol,isopentyl alcohol,isopentanol,3-methylbutanol,1-butanol, 3-methyl,isoamylol,isobutylcarbinol,iso-amylalkohol,isobutyl carbinol
31260
CHEBI:15837
3-methylbutan-1-ol
CC(C)CCO
123-51-3
C5H12O
88.15
PHTQWCKDNZKARW-UHFFFAOYSA-N
3-methyl-1-butanol,isoamyl alcohol,isopentyl alcohol,isopentanol,3-methylbutanol,1-butanol, 3-methyl,isoamylol,isobutylcarbinol,iso-amylalkohol,isobutyl carbinol
31260
CHEBI:15837
3-methylbutan-1-ol
CC(C)CCO
Residue after Evaporation 0.0005 % max.
Infrared Spectrum Authentic
Water 0.005% Max.
Specific Gravity 0.8
Refractive Index 1.4050 to 1.4070
Packaging AcroSeal™ Glass Bottle
Boiling Point 130.0°C
Color Colorless
Quantity 100mL
Density 0.8000g/mL
Flash Point 45°C
Melting Point -70.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2CHCH2CH2OH
Formula Weight 88.15
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3-Methyl-1-butanol
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning