Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 3-Methyl-1-nonyn-3-ol, 97%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Boiling Point | 87°C to 88°C (15mmHg) |
Refractive Index | 1.443 |
Quantity | 25g |
Beilstein | 1750318 |
Formula Weight | 154.25 |
Percent Purity | 97% |
Chemical Name or Material | 3-Methyl-1-nonyn-3-ol |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures