Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Methyl-1-pentanol, 98%

Click to view available options
Quantity:
5g
25g
589-35-5
C6H14O
102.177
MFCD00002937
IWTBVKIGCDZRPL-UHFFFAOYSA-N
3-methyl-1-pentanol,3-methylpentanol,1-pentanol, 3-methyl,2-ethyl-4-butanol,3-ethyl-1-butanol,3-ethylbutanol,3-methyl pentanol,fema no. 3762,3-methyl pentan-1-ol,3-methyl-pentan-1-ol
11508
CHEBI:87381
3-methylpentan-1-ol
CCC(C)CCO
589-35-5
C6H14O
102.177
MFCD00002937
IWTBVKIGCDZRPL-UHFFFAOYSA-N
3-methyl-1-pentanol,3-methylpentanol,1-pentanol, 3-methyl,2-ethyl-4-butanol,3-ethyl-1-butanol,3-ethylbutanol,3-methyl pentanol,fema no. 3762,3-methyl pentan-1-ol,3-methyl-pentan-1-ol
11508
CHEBI:87381
3-methylpentan-1-ol
CCC(C)CCO
Quantity 5g
Flash Point 58°C (136°F)
Boiling Point 151°C to 152°C
UN Number UN1987
Beilstein 1718979
Formula Weight 102.18
Percent Purity 98%
Chemical Name or Material 3-Methyl-1-pentanol

GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

TSCA : TSCA