Alfa Aesar™ 3-Methyl-1-phenyl-2-phospholene 1-oxide, 97%

Manufacturer: Alfa Aesar™ 4472614

Quantity 25g

 View more versions of this product

Catalog No. AA4472614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 707-61-9
    Molecular Formula C11H13OP
    Formula Weight 192.19
    MDL Number MFCD00014518
    Beilstein 609223
    Chemical Name or Material 3-Methyl-1-phenyl-2-phospholene 1-oxide

    Specifications

    Physical Form Lumps
    Formula Weight 192.19
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Molecular Formula C11H13OP
    Odor Unpleasant
    RTECS Number SZ6105100
    MDL Number MFCD00014518
    Health Hazard 1 Warning
    Melting Point 60° to 65°C
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    CAS 707-61-9
    Beilstein 609223
    Color White to cream
    Boiling Point 173° to 174°C (0.7mmHg)
    Flash Point 109°C (228°F)
    Sensitivity Hygroscopic
    Chemical Name or Material 3-Methyl-1-phenyl-2-phospholene 1-oxide
    Assay 97%

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit