Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Methyl-1-phenyl-2-phospholene 1-oxide, 97%

Manufacturer:  Alfa Aesar™ 4472606

Quantity 5g

 View more versions of this product

Catalog No. AA4472606

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

707-61-9
192.19
609223
C11H13OP
MFCD00014518
3-Methyl-1-phenyl-2-phospholene 1-oxide

Specifications

707-61-9
C11H13OP
MFCD00014518
White to cream
173° to 174°C (0.7mmHg)
109°C (228°F)
Hygroscopic
97%
Lumps
192.19
609223
60° to 65°C
5g
Unpleasant
3-Methyl-1-phenyl-2-phospholene 1-oxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : SZ6105100

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit