Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-phenyl-2-pyrazolin-5-one 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
89-25-8
C10H10N2O
174.2
MFCD00003138
QELUYTUMUWHWMC-UHFFFAOYSA-N
edaravone,3-methyl-1-phenyl-2-pyrazolin-5-one,norphenazone,radicut,1-phenyl-3-methyl-5-pyrazolone,developer z,methylphenylpyrazolone,norantipyrine,c.i. developer 1,phenylmethylpyrazolone
4021
CHEBI:31530
5-methyl-2-phenyl-4H-pyrazol-3-one
CC1=NN(C(=O)C1)C2=CC=CC=C2
89-25-8
C10H10N2O
174.2
MFCD00003138
QELUYTUMUWHWMC-UHFFFAOYSA-N
edaravone,3-methyl-1-phenyl-2-pyrazolin-5-one,norphenazone,radicut,1-phenyl-3-methyl-5-pyrazolone,developer z,methylphenylpyrazolone,norantipyrine,c.i. developer 1,phenylmethylpyrazolone
4021
CHEBI:31530
5-methyl-2-phenyl-4H-pyrazol-3-one
CC1=NN(C(=O)C1)C2=CC=CC=C2
Coupling reagent
Color Beige to Yellow
Melting Point 127.0°C to 131.0°C
Boiling Point 287.0°C (265.0 mmHg)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 24, 20
Merck Index 15, 6802
Solubility Information Solubility in water: 3g/L water (20°C). Other solubilities: slightly soluble in alcohol and benzene,insoluble in ether,soluble in acids
Formula Weight 174.2
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 201-891-0

RTECSNumber : UQ9625000

TSCA : TSCA