Alfa Aesar™ 3-Methyl-2(1H)-quinoxalinone, 97%

Manufacturer: Alfa Aesar™ H5173303

Quantity 1g

 View more versions of this product

Catalog No. AAH5173303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14003-34-0
    Molecular Formula C9H8N2O
    Formula Weight 160.18
    MDL Number MFCD00047567
    Chemical Name or Material 3-Methyl-2(1H)-quinoxalinone

    Specifications

    Formula Weight 160.18
    Molecular Formula C9H8N2O
    MDL Number MFCD00047567
    CAS 14003-34-0
    Melting Point 268° to 273°C
    Quantity 1g
    RTECS Number VD3634500
    Chemical Name or Material 3-Methyl-2(1H)-quinoxalinone
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit