Alfa Aesar™ 3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 98+%

Manufacturer: Alfa Aesar™ H5606118

Quantity 50g

 View more versions of this product

Catalog No. AAH5606118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 38894-11-0
    Molecular Formula C8H9N3S·HCl·H2O
    Formula Weight 233.72 (215.70 Anhydrous)
    MDL Number MFCD00149370
    UN Number UN2811
    Beilstein 4722243
    Synonym 2-Hydrazono-3-methylbenzothiazoline hydrochloride monohydrate
    Chemical Name or Material 3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate

    Specifications

    Formula Weight 233.72 (215.70 Anhydrous)
    MDL Number MFCD00149370
    CAS 38894-11-0
    UN Number UN2811
    Melting Point 270° to 274°C
    Quantity 50g
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C8H9N3S·HCl·H2O
    Beilstein 4722243
    Synonym 2-Hydrazono-3-methylbenzothiazoline hydrochloride monohydrate
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    RTECS Number DL7160200
    Chemical Name or Material 3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate
    Assay ≥98%

    Description

    Applications
    It is used as a reagent for the spectrophotometric determination of aliphatic aldehydes and for the determination of hexosamines in glycosaminoglycans. It is Incorporated into a new peroxidase color reaction. It is also a reagent for the spectrophotometric determination of traces of selenium (IV) in environmental samples.

    Solubility
    Soluble in water.

    Notes
    Incompatible with oxidizing agents. Store at room temperature.

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit