Alfa Aesar™ 3-Methyl-2-butanethiol, 95%, remainder mainly 2-methyl-1-butanethiol

Manufacturer: Alfa Aesar™ B2233006

Quantity 5g

 View more versions of this product

Catalog No. AAB2233006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2084-18-6
    Molecular Formula C5H12S
    Formula Weight 104.22
    MDL Number MFCD00039648
    UN Number UN3336
    Chemical Name or Material 3-Methyl-2-butanethiol

    Specifications

    Formula Weight 104.22
    TSCA TSCA
    Molecular Formula C5H12S
    MDL Number MFCD00039648
    UN Number UN3336
    Health Hazard 1 Danger
    Boiling Point 100°C
    Quantity 5g
    Flash Point 3°C (37°F)
    Odor Stench
    Health Hazard 2 GHS H Statement
    H225-H302-H332-H315-H319-H335
    Highly flammable liquid and vapour.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P280g-P305+P351+P338-P403+P233
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store in a well-ventilated place.
    Keep container tightly closed.
    CAS 2084-18-6
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, N.O.S.
    Chemical Name or Material 3-Methyl-2-butanethiol
    Assay 95%, remainder mainly 2-methyl-1-butanethiol

    GHS H Statement
    H225-H302-H332-H315-H319-H335
    Highly flammable liquid and vapour.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280g-P305+P351+P338-P403+P233
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store in a well-ventilated place.
    Keep container tightly closed.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit