Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 3-Methyl-2-butanethiol, 95%, remainder mainly 2-methyl-1-butanethiol
Click to view available options
Quantity:
5g
25g
Specifications
Flash Point | 3°C (37°F) |
Sensitivity | Air Sensitive |
Density | 0.84 |
Refractive Index | 1.444 |
Quantity | 25g |
Odor | Stench |
Boiling Point | 100°C |
UN Number | UN3336 |
Formula Weight | 104.22 |
Percent Purity | 95% |
Show More |
Safety and Handling
GHS H Statement
H225-H302-H332-H315-H319-H335
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, N.O.S.
EINECSNumber : 218-223-9
TSCA : Yes
Recommended Storage : Ambient temperatures