Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-buten-1-ol, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
556-82-1
C5H10O
86.13
MFCD00002916
ASUAYTHWZCLXAN-UHFFFAOYSA-N
3-methyl-2-buten-1-ol,prenol,3,3-dimethylallyl alcohol,2-buten-1-ol, 3-methyl,prenyl alcohol,3-methyl-2-butenol,dimethylallyl alcohol,3-methyl-2-butenyl alcohol,3-methylcrotyl alcohol,3-methyl-2-butene-1-ol
11173
CHEBI:16019
3-methylbut-2-en-1-ol
CC(=CCO)C
556-82-1
C5H10O
86.13
MFCD00002916
ASUAYTHWZCLXAN-UHFFFAOYSA-N
3-methyl-2-buten-1-ol,prenol,3,3-dimethylallyl alcohol,2-buten-1-ol, 3-methyl,prenyl alcohol,3-methyl-2-butenol,dimethylallyl alcohol,3-methyl-2-butenyl alcohol,3-methylcrotyl alcohol,3-methyl-2-butene-1-ol
11173
CHEBI:16019
3-methylbut-2-en-1-ol
CC(=CCO)C
Specific Gravity 0.84
Refractive Index 1.4420 to 1.4440
Boiling Point 140.0°C
Density 0.8400g/mL
Color Colorless to Yellow
Flash Point 43°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula (CH3)2C=CHCH2OH
Beilstein 01, 444
Solubility Information Solubility in water: 170g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 86.13
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3-Methyl-2-buten-1-ol
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta

GHS Signal Word: Danger

EINECSNumber : 209-141-4