Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Methyl-2-butenal, 97%

Manufacturer:  Alfa Aesar™ B2222622

Quantity 100g

 View more versions of this product

Catalog No. AAB2222622


  Check Availability
Add to cart

Specifications

Description

Applications
3-Methyl-2-butenal is used in small quantities as food flavor in alcoholic beverages, chewing gums, confection frosting, frozen dairy, fruit ice, gelatin pudding, hard and soft candy and jam jelly. Used in daily flavor, and cosmetic flavor in perfumes. A starting material for the synthesis of vitamin A.

Solubility
Soluble in water (110g/L).

Notes
Air sensitive. Store away from strong oxidizing agents, air, heat.
Description & Specifications

Chemical Identifiers

107-86-8
(CH3)2C=CHCHO
MFCD00010291
1734740
3,3-Dimethylacrolein; Senecialdehyde
C5H8O
84.12
UN2929
14,8448
3-Methyl-2-butenal

Specifications

107-86-8
(CH3)2C=CHCHO
MFCD00010291
1734740
3,3-Dimethylacrolein; Senecialdehyde
136°C
Characteristic
3-Methyl-2-butenal
C5H8O
84.12
UN2929
14,8448
−20°C
37°C (99°F)
100g
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H331-H318-H226-H302-H315-H317-H335
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P280-P262-P305+P351+P338-P304+P340-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Do not get in eyes, on skin, or on clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.

TSCA : TSCA

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit