Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Methyl-2-butenal, 97%, ACROS Organics™
Click to view available options
Quantity:
5mL
25mL
100mL
Packaging:
Glass bottle
Specifications
Quantity | 5mL |
Flash Point | 33°C |
Color | Colorless to Yellow |
Packaging | Glass bottle |
Specific Gravity | 0.88 |
Infrared Spectrum | Authentic |
Refractive Index | 1.4610 to 1.4630 |
Density | 0.8800g/mL |
Boiling Point | 132.0°C to 133.0°C |
CAS Min % | 96.0 |
Show More |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Danger
EINECSNumber : 203-527-6