Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-butenal, 97%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
Packaging:
Glass bottle
107-86-8
C5H8O
84.12
SEPQTYODOKLVSB-UHFFFAOYSA-N
3-methyl-2-butenal,prenal,3-methylcrotonaldehyde,senecialdehyde,3,3-dimethylacrolein,senecioaldehyde,2-butenal, 3-methyl,beta,beta-dimethylacrolein,crotonaldehyde, 3-methyl,beta-methylcrotonaldehyde
61020
CHEBI:15825
3-methylbut-2-enal
CC(=CC=O)C
107-86-8
C5H8O
84.12
SEPQTYODOKLVSB-UHFFFAOYSA-N
3-methyl-2-butenal,prenal,3-methylcrotonaldehyde,senecialdehyde,3,3-dimethylacrolein,senecioaldehyde,2-butenal, 3-methyl,beta,beta-dimethylacrolein,crotonaldehyde, 3-methyl,beta-methylcrotonaldehyde
61020
CHEBI:15825
3-methylbut-2-enal
CC(=CC=O)C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25mL
Specific Gravity 0.88
Refractive Index 1.4610 to 1.4630
Color Colorless to Yellow
Boiling Point 132.0°C to 133.0°C
Density 0.8800g/mL
Flash Point 33°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)2C=CHCHO
Beilstein 01,IV,3464
Merck Index 15,8587
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol and ether
Formula Weight 84.12
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-Methyl-2-butenal
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta

GHS Signal Word: Danger

EINECSNumber : 203-527-6