Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-butenyl Acetate 98.0+%, TCI America™

Click to view available options
Quantity:
25mL
500mL
1191-16-8
C7H12O2
128.171
MFCD00036569
XXIKYCPRDXIMQM-UHFFFAOYSA-N
prenyl acetate,3,3-dimethylallyl acetate,3-methyl-2-butenyl acetate,isopent-2-enyl acetate,3-methylbut-2-en-1-yl acetate,dimethylallyl acetate,2-buten-1-ol, 3-methyl-, acetate,3-methyl-2-buten-1-ol, acetate,unii-i7kov03hgs,isopentenyl acetate
14489
3-methylbut-2-enyl acetate
CC(=CCOC(=O)C)C
1191-16-8
C7H12O2
128.171
MFCD00036569
XXIKYCPRDXIMQM-UHFFFAOYSA-N
prenyl acetate,3,3-dimethylallyl acetate,3-methyl-2-butenyl acetate,isopent-2-enyl acetate,3-methylbut-2-en-1-yl acetate,dimethylallyl acetate,2-buten-1-ol, 3-methyl-, acetate,3-methyl-2-buten-1-ol, acetate,unii-i7kov03hgs,isopentenyl acetate
14489
3-methylbut-2-enyl acetate
CC(=CCOC(=O)C)C
Boiling Point 128°C
Quantity 500mL
Color Colorless
UN Number 3272
Formula Weight 128.17
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 3-Methyl-2-butenyl Acetate

RTECSNumber : EM9473700

TSCA : Yes