Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-pentene (cis- and trans- mixture) 99.0+%, TCI America™

Click to view available options
Quantity:
1mL
5mL
922-61-2
C6H12
84.162
MFCD00009334
BEQGRRJLJLVQAQ-XQRVVYSFSA-N
643935
(Z)-3-methylpent-2-ene
CCC(=CC)C
922-61-2
C6H12
84.162
MFCD00009334
BEQGRRJLJLVQAQ-XQRVVYSFSA-N
643935
(Z)-3-methylpent-2-ene
CCC(=CC)C
Color Colorless
UN Number 2288
Quantity 5mL
Formula Weight 84.16
Physical Form Clear Liquid at 20°C
Percent Purity ≥99.0% (GC)
Chemical Name or Material 3-Methyl-2-pentene (cis- and trans- mixture)

EINECSNumber : (2)-0031

TSCA : No