Alfa Aesar™ 3-Methyl-3-trimethylsiloxy-1-butyne, 97%

Manufacturer: Alfa Aesar™ 4392204

Quantity 2g

 View more versions of this product

Catalog No. AA4392204

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17869-77-1
    Molecular Formula HC≡CC[OSi(CH3)3](CH3)CH3
    Formula Weight 156.3
    MDL Number MFCD00053867
    UN Number UN1993
    Chemical Name or Material 3-Methyl-3-trimethylsiloxy-1-butyne

    Specifications

    Formula Weight 156.3
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 17869-77-1
    Health Hazard 1 Danger
    Boiling Point 52°C (75mmHg)
    Quantity 2g
    Odor Characteristic
    Physical Form Liquid
    MDL Number MFCD00053867
    Refractive Index 1.4052
    Molecular Formula HC≡CC[OSi(CH3)3](CH3)CH3
    UN Number UN1993
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material 3-Methyl-3-trimethylsiloxy-1-butyne
    Assay 97%

    GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit