Alfa Aesar™ 3-Methyl-4-nitroanisole, 97%

Manufacturer: Alfa Aesar™ B2234822

Quantity 100g

 View more versions of this product

Catalog No. AAB2234822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5367-32-8
    Molecular Formula C8H9NO3
    Formula Weight 167.16
    MDL Number MFCD00007167
    Beilstein 2328600
    Synonym 5-Methoxy-2-nitrotoluene
    Chemical Name or Material 3-Methyl-4-nitroanisole

    Specifications

    Formula Weight 167.16
    Molecular Formula C8H9NO3
    MDL Number MFCD00007167
    CAS 5367-32-8
    Beilstein 2328600
    Synonym 5-Methoxy-2-nitrotoluene
    Health Hazard 1 Danger
    Melting Point 48° to 50°C
    Quantity 100g
    Flash Point >110°C (230°F)
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 3-Methyl-4-nitroanisole
    Assay 97%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit