Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Methyl-4-nitrobenzamide, 97%

Click to view available options
Quantity:
1g
5g
25g
99584-85-7
C8H8N2O3
180.163
MFCD00031468
BCAAGQMIPKPDDX-UHFFFAOYSA-N
3-methyl-4-nitro-benzamide,3-methyl-4-nitro benzamide,#,benzamide, 3-methyl-4-nitro,3-methyl-4-nitrobenzoic acid amide
595900
3-methyl-4-nitrobenzamide
CC1=C(C=CC(=C1)C(=O)N)[N+](=O)[O-]
99584-85-7
C8H8N2O3
180.163
MFCD00031468
BCAAGQMIPKPDDX-UHFFFAOYSA-N
3-methyl-4-nitro-benzamide,3-methyl-4-nitro benzamide,#,benzamide, 3-methyl-4-nitro,3-methyl-4-nitrobenzoic acid amide
595900
3-methyl-4-nitrobenzamide
CC1=C(C=CC(=C1)C(=O)N)[N+](=O)[O-]
Applications
3-Methyl-4-nitrobenzamide is used as synthetic organic intermediates.

Solubility
Soluble in alcohol, slightly soluble in hot water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Quantity 5g
Melting Point 158°C to 160°C
Beilstein 2264048
Solubility Information Soluble in alcohol,slightly soluble in hot water.
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-nitrobenzamide

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures