Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Methyl-4-nitrobenzamide, 97%

Click to view available options
Quantity:
1g
5g
25g
99584-85-7
C8H8N2O3
180.163
MFCD00031468
BCAAGQMIPKPDDX-UHFFFAOYSA-N
3-methyl-4-nitro-benzamide,3-methyl-4-nitro benzamide,#,benzamide, 3-methyl-4-nitro,3-methyl-4-nitrobenzoic acid amide
595900
3-methyl-4-nitrobenzamide
CC1=C(C=CC(=C1)C(=O)N)[N+](=O)[O-]
99584-85-7
C8H8N2O3
180.163
MFCD00031468
BCAAGQMIPKPDDX-UHFFFAOYSA-N
3-methyl-4-nitro-benzamide,3-methyl-4-nitro benzamide,#,benzamide, 3-methyl-4-nitro,3-methyl-4-nitrobenzoic acid amide
595900
3-methyl-4-nitrobenzamide
CC1=C(C=CC(=C1)C(=O)N)[N+](=O)[O-]
Applications
3-Methyl-4-nitrobenzamide is used as synthetic organic intermediates.

Solubility
Soluble in alcohol, slightly soluble in hot water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Quantity 25g
Melting Point 158°C to 160°C
Beilstein 2264048
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-nitrobenzamide

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger