Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-4-nitrobenzoic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
3113-71-1
C8H7NO4
181.15
MFCD00007168
XDTTUTIFWDAMIX-UHFFFAOYSA-N
3-methyl-4-nitrobenzoic acid,4-nitro-m-toluic acid,benzoic acid, 3-methyl-4-nitro,m-toluic acid, 4-nitro,unii-elp258jxh0,3-methyl-4-nitrobenzoicacid,3-methyl-4-nitro-benzoic acid,elp258jxh0,benzoic acid,3-methyl-4-nitro,3-methyl-4-nitro benzoic acid
18370
CC1=C(C=CC(=C1)C(=O)O)[N+](=O)[O-]
3113-71-1
C8H7NO4
181.15
MFCD00007168
XDTTUTIFWDAMIX-UHFFFAOYSA-N
3-methyl-4-nitrobenzoic acid,4-nitro-m-toluic acid,benzoic acid, 3-methyl-4-nitro,m-toluic acid, 4-nitro,unii-elp258jxh0,3-methyl-4-nitrobenzoicacid,3-methyl-4-nitro-benzoic acid,elp258jxh0,benzoic acid,3-methyl-4-nitro,3-methyl-4-nitro benzoic acid
18370
CC1=C(C=CC(=C1)C(=O)O)[N+](=O)[O-]
Packaging Plastic bottle
Infrared Spectrum Authentic
Color Yellow
Melting Point 216°C to 219°C
Quantity 500g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3C6H3(NO2)CO2H
Beilstein 09, 481
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in most organic solvents
Formula Weight 181.15
Physical Form Amorphous Powder
Percent Purity ≥98.5%
Chemical Name or Material 3-Methyl-4-nitrobenzoic acid
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning