Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-5-(5-methylisoxazol-3-yl)isoxazole-4-carboxylic acid, 97%, Maybridge

Click to view available options
Quantity:
1g
849066-63-3
C9H8N2O4
208.173
MFCD01880302
ASBPDKJMOPNMLF-UHFFFAOYSA-N
3-methyl-5-5-methylisoxazol-3-yl isoxazole-4-carboxylic acid,3-methyl-5-5-methyl-1,2-oxazol-3-yl-1,2-oxazole-4-carboxylic acid,3,5'-biisoxazole-4'-carboxylicacid, 3',5-dimethyl,maybridge3_007228,3',5-dimethyl-3,5'-biisoxazole-4'-carboxylic acid,3-methyl-5-5-methylisoxazol-3-yl-4-carboxylic acid
2744621
3-methyl-5-(5-methyl-1,2-oxazol-3-yl)-1,2-oxazole-4-carboxylic acid
CC1=CC(=NO1)C2=C(C(=NO2)C)C(=O)O
849066-63-3
C9H8N2O4
208.173
MFCD01880302
ASBPDKJMOPNMLF-UHFFFAOYSA-N
3-methyl-5-5-methylisoxazol-3-yl isoxazole-4-carboxylic acid,3-methyl-5-5-methyl-1,2-oxazol-3-yl-1,2-oxazole-4-carboxylic acid,3,5'-biisoxazole-4'-carboxylicacid, 3',5-dimethyl,maybridge3_007228,3',5-dimethyl-3,5'-biisoxazole-4'-carboxylic acid,3-methyl-5-5-methylisoxazol-3-yl-4-carboxylic acid
2744621
3-methyl-5-(5-methyl-1,2-oxazol-3-yl)-1,2-oxazole-4-carboxylic acid
CC1=CC(=NO1)C2=C(C(=NO2)C)C(=O)O
Packaging Amber Glass Bottle
Quantity 1g
CAS Min % 97.0
Assay Percent Range 97%
Formula Weight 208.17
Percent Purity 97%
Chemical Name or Material 3-Methyl-5-(5-methylisoxazol-3-yl)isoxazole-4-carboxylic acid

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.