Alfa Aesar™ 3-Methyl-5-(trifluoromethyl)benzyl bromide, 97%

Manufacturer: Alfa Aesar™ H3352003

Quantity 1g

 View more versions of this product

Catalog No. AAH3352003

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C9H8BrF3
    Formula Weight 253.06
    MDL Number MFCD01631610
    UN Number UN3265
    Chemical Name or Material 3-Methyl-5-(trifluoromethyl)benzyl bromide

    Specifications

    Formula Weight 253.06
    Molecular Formula C9H8BrF3
    MDL Number MFCD01631610
    UN Number UN3265
    Quantity 1g
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Chemical Name or Material 3-Methyl-5-(trifluoromethyl)benzyl bromide
    Assay 97%

    Description

    Applications
    A series of novel five-membered urea derivatives as potent NK1 receptor antagonists is described. The effects of substitution of a 4-fluoro group at the phenyl ring and the introduction of an α-methyl group at the benzylic position to improve potency was found. 3,5-Bis(trifluoromethyl)benzyl bromide has been used as derivatization reagent in detection of uracil in DNA by GC and negative chemical ionization mass spectrometry.

    Notes
    Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit