Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Methyl-5-(trifluoromethyl)benzyl bromide, 97%

Manufacturer:  Alfa Aesar™ H33520MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH33520MD


  Check Availability
Add to cart

Specifications

Description

Applications
A series of novel five-membered urea derivatives as potent NK1 receptor antagonists is described. The effects of substitution of a 4-fluoro group at the phenyl ring and the introduction of an α-methyl group at the benzylic position to improve potency was found. 3,5-Bis(trifluoromethyl)benzyl bromide has been used as derivatization reagent in detection of uracil in DNA by GC and negative chemical ionization mass spectrometry.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water.
Description & Specifications

Chemical Identifiers

C9H8BrF3
MFCD01631610
3-Methyl-5-(trifluoromethyl)benzyl bromide
253.06
UN3265

Specifications

C9H8BrF3
MFCD01631610
250mg
97%
253.06
UN3265
3-Methyl-5-(trifluoromethyl)benzyl bromide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit