Alfa Aesar™ 3-Methyl-beta-styrylboronic acid pinacol ester, 97%

Manufacturer: Alfa Aesar™ H3007306

Quantity 5g

 View more versions of this product

Catalog No. AAH3007306

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C15H21BO2
    Formula Weight 244.14
    MDL Number MFCD12546181
    Synonym 4,4,5,5-Tetramethyl-2-(trans-2-m-tolylvinyl)[1,3,2]dioxaborolane
    Chemical Name or Material 3-Methyl-beta-styrylboronic acid pinacol ester

    Specifications

    Formula Weight 244.14
    Synonym 4,4,5,5-Tetramethyl-2-(trans-2-m-tolylvinyl)[1,3,2]dioxaborolane
    Boiling Point 114°C (0.3mmHg)
    Quantity 5g
    Molecular Formula C15H21BO2
    MDL Number MFCD12546181
    Chemical Name or Material 3-Methyl-beta-styrylboronic acid pinacol ester
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit