Alfa Aesar™ 3-Methyl-D-phenylalanine, 98%

Manufacturer: Alfa Aesar™ H2710803

Quantity 1g

 View more versions of this product

Catalog No. AAH2710803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 114926-39-5
    Molecular Formula C10H13NO2
    Formula Weight 179.22
    MDL Number MFCD01860871
    Synonym H-D-Phe(3-Me)-OH
    Chemical Name or Material 3-Methyl-D-phenylalanine

    Specifications

    Formula Weight 179.22
    Molecular Formula C10H13NO2
    CAS 114926-39-5
    Synonym H-D-Phe(3-Me)-OH
    Quantity 1g
    MDL Number MFCD01860871
    Chemical Name or Material 3-Methyl-D-phenylalanine
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit