Alfa Aesar™ 3-Methyl(thiobenzamide), 97%

Manufacturer: Alfa Aesar™ H2646803

Quantity 1g

 View more versions of this product

Catalog No. AAH2646803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2362-63-2
    Molecular Formula C8H9NS
    Formula Weight 151.23
    MDL Number MFCD01314035
    Synonym 3-Methylbenzene-1-carbothioamide
    Chemical Name or Material 3-Methyl(thiobenzamide)

    Specifications

    Formula Weight 151.23
    Molecular Formula C8H9NS
    MDL Number MFCD01314035
    Synonym 3-Methylbenzene-1-carbothioamide
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P280f
    Wear protective clothing.
    Melting Point 87° to 89°C
    Quantity 1g
    CAS 2362-63-2
    Chemical Name or Material 3-Methyl(thiobenzamide)
    Assay 97%

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P280f
    Wear protective clothing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit