Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylamino-1,2-propanediol, 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
Packaging:
Glass bottle
40137-22-2
C4H11NO2
105.137
MFCD00082732
WOMTYMDHLQTCHY-UHFFFAOYSA-N
3-methylamino-1,2-propanediol,3-methylamino propane-1,2-diol,1,2-propanediol, 3-methylamino,acmc-209jb1,ksc495i3p,3-methylamino-1,2-propandiol,n-methylamino-2,3-propanediol,3-methylamino-propan-1,2-diol,3-methylamino-propane-1,2-diol
2734122
3-(methylamino)propane-1,2-diol
CNCC(CO)O
40137-22-2
C4H11NO2
105.137
MFCD00082732
WOMTYMDHLQTCHY-UHFFFAOYSA-N
3-methylamino-1,2-propanediol,3-methylamino propane-1,2-diol,1,2-propanediol, 3-methylamino,acmc-209jb1,ksc495i3p,3-methylamino-1,2-propandiol,n-methylamino-2,3-propanediol,3-methylamino-propan-1,2-diol,3-methylamino-propane-1,2-diol
2734122
3-(methylamino)propane-1,2-diol
CNCC(CO)O
Infrared Spectrum Authentic
Refractive Index 1.4740 to 1.4780 (20°C, 589nm)
Quantity 25mL
Density 1.09g/mL
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula H3CNHCHCH(OH)CHOH
Formula Weight 105.14
Physical Form Viscous Liquid
Percent Purity ≥97.5%
Chemical Name or Material 3-Methylamino-1, 2-propanediol, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 254-809-0

TSCA : TSCA