Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Methylbenzene-1,2-diamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Linear Formula | CH3C6H3(NH2)2 |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Formula Weight | 122.17 |
Percent Purity | 97% |
Chemical Name or Material | 3-Methylbenzene-1, 2-diamine |