Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Methylbenzeneboronic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
17933-03-8
C7H9BO2
135.957
MFCD00040198
BJQCPCFFYBKRLM-UHFFFAOYSA-N
3-tolylboronic acid,m-tolylboronic acid,3-methylphenyl boronic acid,3-methylbenzeneboronic acid,3-methyl phenyl boronic acid,3-tolyboronic acid,3-methylphenyl boranediol,m-methylphenylboronic acid,m-tolyl boronic acid
2733950
(3-methylphenyl)boronic acid
B(C1=CC(=CC=C1)C)(O)O
17933-03-8
C7H9BO2
135.957
MFCD00040198
BJQCPCFFYBKRLM-UHFFFAOYSA-N
3-tolylboronic acid,m-tolylboronic acid,3-methylphenyl boronic acid,3-methylbenzeneboronic acid,3-methyl phenyl boronic acid,3-tolyboronic acid,3-methylphenyl boranediol,m-methylphenylboronic acid,m-tolyl boronic acid
2733950
(3-methylphenyl)boronic acid
B(C1=CC(=CC=C1)C)(O)O
Melting Point ~160°C to 162°C
Quantity 25g
Beilstein 2935968
Formula Weight 135.96
Percent Purity 97%
Chemical Name or Material 3-Methylbenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : ED7788888